VWR® Heating Blocks for Block Heaters
Supplier: VWR Collection
13259-256 13259-297
VWRI460-3236EA 268 SGD
VWRI460-3236 VWRI460-3237 VWRI460-3238 VWRI460-3239 VWRI460-3224 VWRI460-3225 VWRI460-3226 VWRU13259-256 VWRU13259-297
VWR® Heating Blocks for Block Heaters
Block Heaters Blocks
Solid blocks: For use as a low temperature hotplate or for custom drilling.
  • For use as a low-temperature hotplate or for custom drilling
Blocks for microtitre plates: Recessed well for better stability; flat surface is good for flat and round bottom plates. Fits 2/4/6 block heaters.
Blocks for PCR - plates, tubes, strips: Blocks for PCR - plates, tubes, strips: Tapered tube wells for 0,2 ml tubes. Spaced for easy access and removal.
Blocks for cuvettes: Two parallel slots fit six cuvettes in each slot, side by side for excellent stability and heat transfer.
Order Now
SPECIFICATIONS
Environmentally Preferable