VWR®, Remote Temperature Probes for VWR Chillers
Supplier: VWR Collection
VWRI461-0101EA 1160 SGD
VWRI461-0101 VWRI461-0102 VWRI461-0103
VWR®, Remote Temperature Probes for VWR Chillers
Temperature Probes
Remote temperature probes for circulator use with a chiller. Allows points of control to be switched from inside a chiller to a remote location. Compensates for heat loss through tubing between the circulator and the control point. Platinum RTD sensor with stainless steel sheath. Rubber coupling with 25,4 mm Ø, connects sheath to flexible cable. Probe cable ends in a DB9 female plug.
Lengthר: 152×4 mm
Order Now
SPECIFICATIONS
Environmentally Preferable