VWR®, Tubing, High Temperature for Circulators
Supplier: VWR Collection
VWRI461-0106EA 851 SGD
VWRI461-0106
VWR®, Tubing, High Temperature for Circulators
Tubing Transfer Tubing
PTFE lined, stainless steel braid.
Flexible PTFE lined, stainless steel over braided tubing with 6,3 mm NPT quick connectors at both ends. Maximum temperature is 300 °C.
Length (mm): 930
Order Now
SPECIFICATIONS
Environmentally Preferable