VWR®, Coliform Bath with MX Temperature Controller
Supplier: VWR Collection
VWRI462-0214EA 10340 SGD
VWRI462-0214
VWR®, Coliform Bath with MX Temperature Controller
Baths Coliform Baths
Stainless steel bath with user friendly MX temperature controller, has three control buttons and on-screen prompts. Specifically designed for production of microbiological culture media. The controller bridge, made from chemical resistant DuraTop™, is easily removed to allow cleaning of the tank. The bath has an opening with lid for optional cooling coil, which is ideal for set points near ambient temperature.
  • Large EasyView™ LCD display with alarm or fault icons plus English prompts and menus
  • User-adjustable high and low temperature limits and low liquid
More Product Information