VWR®, Refrigerated Circulators
Supplier: VWR Collection
89202-974 89202-976
VWRI462-0238EA 39660 SGD
VWRI462-0238 VWRI462-0237 VWRI462-0236 VWRI462-0235 VWRI462-0234 VWRI462-0233 VWRI462-0232 VWRI462-0230 VWRI462-0228 VWRI462-0227 VWRI462-0226 VWRI462-0224 VWRI462-0239 VWRI462-0231 VWRU89202-974 VWRU89202-976
VWR®, Refrigerated Circulators
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Stainless steel circulating baths with choice of Advanced Programmable or Advanced Digital temperature controller. Both are easy to navigate with large, intuitive displays and multiple communication options, including USB-A and B, RS232/485, Ethernet and external temperature probe. The reservoir drain can be accessed by removing the front panel. All models feature user-adjustable, high temperature safety cut-off points, as well as over-temperature protection. The surface is cool to the touch, even when operating at high temperatures.
  • Working temperatures from −40 to +200 °C with
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable