VWR®, Tube Racks for VWB Series Baths
Supplier: VWR Collection
VWRI462-0253EA 233 SGD
VWRI462-0253 VWRI462-0254 VWRI462-0255 VWRI462-0256 VWRI462-0257 VWRI462-0258 VWRI462-0252
VWR®, Tube Racks for VWB Series Baths
Racks Tube Racks
Tube racks suitable for: VWB 6, VWB 12, VWB 18, VWB 26, maximum 1, 4, 7, 7 racks per bath respectively.
Order Now
SPECIFICATIONS
Environmentally Preferable