VWR®, Accessories for Circulators
Supplier: VWR Collection
VWRI462-0271EA 1230 SGD
VWRI462-0271 VWRI462-0272 VWRI462-0273 VWRI462-0274
VWR®, Accessories for Circulators
Circulating Thermostats Open Circulator Systems
Stainless steel probes suitable for used with any instrument that has an Advanced Digital (AD) or Advanced Programmable (AP) controller.
Probe measurements: 152 mm length, 4,7 mm Ø.
Order Now
SPECIFICATIONS
Environmentally Preferable