VWR®, Tubing, Viton®, for Circulators
Supplier: VWR Collection
VWRI462-0300EA 859 SGD
VWRI462-0300
VWR®, Tubing, Viton®, for Circulators
Tubing Transfer Tubing
Fluoroelastomer, black.
  • Excellent resistance to corrosive chemicals, oils, fuels and solvents
  • Resistant to ozone, sunlight and weathering
  • Tear resistant and durable
Temperature resistance: −40 to +150 °C.