VWR® VWB2, Water Baths
Supplier: VWR Collection
VWRI462-0554EA 1480 SGD
VWRI462-0554 VWRI462-0555 VWRI462-0556 VWRI462-0557 VWRI462-0558 VWRI462-0559
VWR® VWB2, Water Baths
Baths Water Baths, Unstirred
Unstirred water baths with double walled, stainless steel (inner) and coated steel sheet (outer) housing. All baths except the 2 litre models feature a drain tap for convenient emptying and are suitable for use with water or alternatively with heat transfer beads. Baths with a capacity of 5 litres or more have a heat transfer bead mode which is activated through the main menu. A drain stopper (plug) is supplied as on optional accessory to ensure heat transfer beads do not get stuck in the drain outlet. Note: Baths are classified Safety Class 1, allowing unattended operation when used withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable