VWR® VENTI-Line® Prime, Drying Ovens with Forced Convection
Supplier: VWR Collection
VWRI466-0603EA 3710 SGD
VWRI466-0603 VWRI466-0604 VWRI466-0605
VWR® VENTI-Line® Prime, Drying Ovens with Forced Convection
Ovens Standard Ovens
VENTI-Line® drying ovens are designed for all standard drying and tempering tasks as well as sterilising glassware. They have a temperature range between 50 to 300 °C and are intended for use in biological, chemical, medical, pharmaceutical and physical laboratories.
  • Multifunctional microprocessor controller with 4,3" full colour touch screen
  • Five user programs memory
  • Five segment temperature-time profile, quick change of set temperature and set time during a running program
  • Scheduled program start (set of date and time) and 'Loop' function up to 255 times
More Product Information