VWR® Desiccators
Supplier: VWR Collection
VWRI467-0071EA 298 SGD
VWRI467-0071 VWRI467-0069 VWRI467-0070 VWRI467-0072 VWRI467-0073
VWR® Desiccators
Desiccators Desiccators, Glass
Borosilicate 3.3 glass.
  • Precisely ground flat flanges and interchangeable knobbed lids
  • Base and lid with flat flange ground to fit
  • High mechanical, thermal and chemical resistance
These desiccators are ideal for drying, drying processes and protecting samples from the effects of air and moisture.
Certifications: DIN 12491