VWR® CSSXSS, Cryogenic Storage Vessels
Supplier: VWR Collection
VWRI478-0285EA 1780 SGD
VWRI478-0285
VWR® CSSXSS, Cryogenic Storage Vessels
Dewars and Cryogenic Vessels
CSS series tanks offer storage for small to medium capacity applications. The XSS series offer similar benefits and extended storage capacity. The tanks are manufactured from aluminium alloy with fibre glass/epoxy neck. High vacuum, multi-layered super insulation limits nitrogen evaporation.
  • Extremely low liquid nitrogen evaporation
  • Lid is lockable with padlock (not included)
  • Vessels are equipped with stainless steel canisters with hook for easy retrieval from the vessel
The samples are placed in stainless steel canisters directly in the liquidMore Product Information