VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
VWRI479-7019EA 3.8 SGD
VWRI479-7019 VWRI479-7020 VWRI479-7021 VWRI479-7022 VWRI479-7023 VWRI479-7024 VWRI479-7025 VWRI479-7026 VWRI479-7027 VWRI479-7028 VWRI479-7029 VWRI479-7030 VWRI479-7031 VWRI479-7032 VWRI479-7033 VWRI479-7034 VWRI479-7035
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Boxes Cryoboxes
Cardboard, coated.
  • Waterproof coating
  • Freezable to –140 °C
  • Available with or without dividers
  • In different heights and five colours
For storage of cryovials at low temperatures.
Order Now
SPECIFICATIONS
Environmentally Preferable