VWR®, Test Sieves, 200×32 mm
Supplier: VWR Collection
VWRI510-0608EA 2090 SGD
VWRI510-0608 VWRI510-0609 VWRI510-0610 VWRI510-0611 VWRI510-0612 VWRI510-0613 VWRI510-0614 VWRI510-0615 VWRI510-0616 VWRI510-0617 VWRI510-0618 VWRI510-0619 VWRI510-0620 VWRI510-0621 VWRI510-0622 VWRI510-0623 VWRI510-0624 VWRI510-0625 VWRI510-0626 VWRI510-0627 VWRI510-0628 VWRI510-0629 VWRI510-0630 VWRI510-0631 VWRI510-0632 VWRI510-0633 VWRI510-0634 VWRI510-0635 VWRI510-0636 VWRI510-0637 VWRI510-0638 VWRI510-0639 VWRI510-0640 VWRI510-0641 VWRI510-0642 VWRI510-0643 VWRI510-0644 VWRI510-0645 VWRI510-0646 VWRI510-0647 VWRI510-0648 VWRI510-0649 VWRI510-0650 VWRI510-0651 VWRI510-0652 VWRI510-0653 VWRI510-0654 VWRI510-0655 VWRI510-0656 VWRI510-0657 VWRI510-0658 VWRI510-0659 VWRI510-0660 VWRI510-0661 VWRI510-0662 VWRI510-0663 VWRI510-0664 VWRI510-0665 VWRI510-0666 VWRI510-0667 VWRI510-0668 VWRI510-0669 VWRI510-0670 VWRI510-0671 VWRI510-0672 VWRI510-0673 VWRI510-0674 VWRI510-0675 VWRI510-0676 VWRI510-0677 VWRI510-0678
VWR®, Test Sieves, 200×32 mm
Sieves Test Sieves
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.