VWR®, Test Sieves, 200×50 mm
Supplier: VWR Collection
VWRI510-0700EA 2040 SGD
VWRI510-0700 VWRI510-0701 VWRI510-0702 VWRI510-0703 VWRI510-0704 VWRI510-0705 VWRI510-0706 VWRI510-0707 VWRI510-0708 VWRI510-0709 VWRI510-0710 VWRI510-0711 VWRI510-0712 VWRI510-0713 VWRI510-0714 VWRI510-0715 VWRI510-0716 VWRI510-0717 VWRI510-0718 VWRI510-0719 VWRI510-0720 VWRI510-0721 VWRI510-0722 VWRI510-0723 VWRI510-0724 VWRI510-0725 VWRI510-0726 VWRI510-0727 VWRI510-0728 VWRI510-0729 VWRI510-0730 VWRI510-0731 VWRI510-0732 VWRI510-0733 VWRI510-0734 VWRI510-0735 VWRI510-0736 VWRI510-0737 VWRI510-0738 VWRI510-0739 VWRI510-0740 VWRI510-0741 VWRI510-0742 VWRI510-0743 VWRI510-0744 VWRI510-0745 VWRI510-0746 VWRI510-0747 VWRI510-0748 VWRI510-0749 VWRI510-0750 VWRI510-0751 VWRI510-0752 VWRI510-0753 VWRI510-0754 VWRI510-0755 VWRI510-0756 VWRI510-0757 VWRI510-0758 VWRI510-0759 VWRI510-0760 VWRI510-0761 VWRI510-0762 VWRI510-0763 VWRI510-0764 VWRI510-0765 VWRI510-0766 VWRI510-0767 VWRI510-0768 VWRI510-0769 VWRI510-0770 VWRI510-0771 VWRI510-0772 VWRI510-0773 VWRI510-0774 VWRI510-0775 VWRI510-0776 VWRI510-0777 VWRI510-0778 VWRI510-0779 VWRI510-0780 VWRI510-0781 VWRI510-0782 VWRI510-0783 VWRI510-0784 VWRI510-0785 VWRI510-0786 VWRI510-0787 VWRI510-0788 VWRI510-0789 VWRI510-0790 VWRI510-0791 VWRI510-0792 VWRI510-0793 VWRI510-0794 VWRI510-0795 VWRI510-0796 VWRI510-0797 VWRI510-0798
VWR®, Test Sieves, 200×50 mm
Sieves Test Sieves
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.