VWR®, Test Sieves, 300×60 mm
Supplier: VWR Collection
VWRI510-2697EA 664 SGD
VWRI510-2697 VWRI510-2698 VWRI510-2699 VWRI510-2700 VWRI510-2701 VWRI510-2702 VWRI510-2703 VWRI510-2704 VWRI510-2705 VWRI510-2706 VWRI510-2707 VWRI510-2708 VWRI510-2709 VWRI510-2710 VWRI510-2711 VWRI510-2712 VWRI510-2713 VWRI510-2714 VWRI510-2715 VWRI510-2716 VWRI510-2717 VWRI510-2718 VWRI510-2719 VWRI510-2720 VWRI510-2721 VWRI510-2722 VWRI510-2723 VWRI510-2724 VWRI510-2725 VWRI510-2726 VWRI510-2727 VWRI510-2728 VWRI510-2729 VWRI510-2730 VWRI510-2731 VWRI510-2732 VWRI510-2733 VWRI510-2734 VWRI510-2735 VWRI510-2736 VWRI510-2737 VWRI510-2738 VWRI510-2739 VWRI510-2740 VWRI510-2741 VWRI510-2742 VWRI510-2743 VWRI510-2744 VWRI510-2745 VWRI510-2746 VWRI510-2747 VWRI510-2748 VWRI510-2749 VWRI510-2750 VWRI510-2751 VWRI510-2752 VWRI510-2753 VWRI510-2754 VWRI510-2755 VWRI510-2756 VWRI510-2757 VWRI510-2758 VWRI510-2759 VWRI510-2760 VWRI510-2761 VWRI510-2762 VWRI510-2763 VWRI510-2764 VWRI510-2765 VWRI510-2766 VWRI510-2767 VWRI510-2768 VWRI510-2769 VWRI510-2770 VWRI510-2771 VWRI510-2772 VWRI510-2773 VWRI510-2774 VWRI510-2775 VWRI510-2776 VWRI510-2777 VWRI510-2778 VWRI510-2779 VWRI510-2780 VWRI510-2781 VWRI510-2782 VWRI510-2783 VWRI510-2784 VWRI510-2785 VWRI510-2786 VWRI510-2787 VWRI510-2788
VWR®, Test Sieves, 300×60 mm
Sieves Test Sieves
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.