VWR®, Test Sieves, 350×60 mm
Supplier: VWR Collection
VWRI510-3338EA 796 SGD
VWRI510-3338 VWRI510-3339 VWRI510-3340 VWRI510-3341 VWRI510-3342 VWRI510-3343 VWRI510-3349 VWRI510-3344 VWRI510-3345 VWRI510-3346 VWRI510-3347 VWRI510-3348 VWRI510-3350 VWRI510-3351 VWRI510-3352 VWRI510-3353 VWRI510-3354 VWRI510-3355 VWRI510-3357 VWRI510-3358 VWRI510-3359 VWRI510-3360 VWRI510-3361 VWRI510-3362 VWRI510-3363 VWRI510-3364 VWRI510-3356 VWRI510-3365 VWRI510-3366 VWRI510-3367 VWRI510-3368 VWRI510-3369 VWRI510-3370 VWRI510-3371 VWRI510-3372 VWRI510-3373 VWRI510-3374 VWRI510-3375 VWRI510-3376 VWRI510-3377 VWRI510-3378 VWRI510-3379 VWRI510-3380 VWRI510-3381 VWRI510-3382 VWRI510-3383 VWRI510-3384 VWRI510-3385 VWRI510-3386 VWRI510-3387 VWRI510-3388 VWRI510-3389 VWRI510-3390 VWRI510-3391 VWRI510-3392 VWRI510-3393 VWRI510-3394 VWRI510-3395 VWRI510-3396 VWRI510-3397 VWRI510-3398 VWRI510-3399 VWRI510-3400 VWRI510-3401 VWRI510-3402 VWRI510-3403 VWRI510-3404 VWRI510-3405 VWRI510-3406 VWRI510-3407 VWRI510-3408 VWRI510-3409 VWRI510-3410 VWRI510-3411 VWRI510-3412 VWRI510-3413 VWRI510-3414 VWRI510-3415 VWRI510-3416 VWRI510-3417 VWRI510-3418 VWRI510-3419 VWRI510-3420 VWRI510-3421 VWRI510-3422 VWRI510-3423 VWRI510-3424 VWRI510-3425 VWRI510-3426 VWRI510-3427 VWRI510-3428 VWRI510-3429
VWR®, Test Sieves, 350×60 mm
Sieves Test Sieves
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.