VWR® Tube Top Vacuum Filtration Systems
Supplier: VWR Collection
VWRI514-1068EA 181 SGD
VWRI514-1068 VWRI514-1069 VWRI514-1070 VWRI514-1071
VWR® Tube Top Vacuum Filtration Systems
Filters Bottle Top Filters
These units are ideal for minimising unnecessary transfers by filtering directly into the centrifuge tube.
  • Sterilised by electron beam irradiation
  • Non pyrogenic
PES membrane: Low protein binding and low extractables. Ideal for tissue culture applications.
Delivery information: Each PP centrifuge tube is supplied with an individually wrapped cap for storage. Each individual unit is lot-numbered for easy identification and tracking.
Packaging: Packaged in resealable bag. Centrifuge tube cap is individually wrapped.
Caution: Funnel/tube size.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M