VWR® Phase Separating Paper, Grade 511
Supplier: VWR Collection
VWRI516-0313EA 119 SGD
VWRI516-0313 VWRI516-0314 VWRI516-0315 VWRI516-0311 VWRI516-0312
VWR® Phase Separating Paper, Grade 511
Filters Filter Papers Phase Separation Paper
Hydrophobic filter paper used for separation of aqueous solutions from organic solutions.
  • Usable as a disposable separating funnel