VWR® Micro Star 12, Microcentrifuge
Supplier: VWR Collection
VWRI521-1651EA 2560 SGD
VWRI521-1651
VWR® Micro Star 12, Microcentrifuge
Centrifuges Microcentrifuges
Safe and easy to use centrifuge, delivering optimum performance in a compact unit.
  • Compact design with small footprint
  • Rapid acceleration and deceleration together with quiet operation characteristics
  • Alarms for over-speed and over-heat
  • Autoclavable rotor for 12×1,5/2,0 ml tubes includes biosafety rotor lid
All operating parameters, including rpm, RCF and remaining time, are set and displayed digitally. It has an automatic lid release, pulse-spin function for fast, convenient quick spins, and a unique internal airflow design to reduce the heating upMore Product Information