VWR®, Centrifuge Tubes
Supplier: VWR Collection
VWRI525-0178EA 29.9 SGD
VWRI525-0178 VWRI525-0179 VWRI525-0180 VWRI525-0181 VWRI525-0182
VWR®, Centrifuge Tubes
Tubes Centrifuge Tubes
PTFE, round bottom, smooth surface, without screw cap.
  • Inert
  • Withstand forces up to 12500×g
  • Excellent chemical resistance
Suitable for use at high and low temperatures (–200 to +280 °C).
Order Now
SPECIFICATIONS
Environmentally Preferable