VWR®, Centrifuge Tubes, PTFE
Supplier: VWR Collection
VWRI525-0187EA 37.9 SGD
VWRI525-0187 VWRI525-0188 VWRI525-0189 VWRI525-0190 VWRI525-0191 VWRI525-0183 VWRI525-0184 VWRI525-0185 VWRI525-0186
VWR®, Centrifuge Tubes, PTFE
Tubes Centrifuge Tubes
PTFE, smooth surface, with or without screw cap.
  • Inert
  • Withstand forces up to 12500×g
  • Excellent chemical resistance
Suitable for use at high and low temperatures (–200 to +280 °C).
Order Now
SPECIFICATIONS
Environmentally Preferable