VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWRI525-0606EA 124 SGD
VWRI525-0606 VWRI525-0611 VWRI525-0607 VWRI525-0608 VWRI525-0609 VWRI525-0610 VWRI525-0605 VWRI525-0604 VWRI525-0612
VWR®, Centrifuge Tubes, Conical Bottom
Tubes Centrifuge Tubes
PP, conical, with white HDPE flat cap or plug cap.
  • Certified non cytotoxic, nuclease-free and pyrogen-free
  • Caps available in two styles: Flat cap with a smooth surface for additional labelling options, or plug style cap for a secure seal when using shakers and horizontal incubators
  • Engraved graduation at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • Can be used for storing and freezing samples at temperatures down to –80 °C
  • Sustainable design based on virgin resins
  • During the freezing process, it is better to use
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M