VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
VWRI525-0633EA 285 SGD
VWRI525-0633 VWRI525-0635 VWRI525-0634
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Tubes Centrifuge Tubes
Manufactured of high-quality USP Class 6 gamma resistent clear polypropylene with HDPE screw cap.
  • Longer length screw caps
  • Easy-to-read black graduations are accurate to ±2%
  • Large unerasable frosting white printed writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Autoclavable at 121 °C and freezable to –80 °C
  • Leak-Proof
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
These disposable freestanding tubes feature black printedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M