VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Supplier: VWR Collection
VWRI525-0636EA 281 SGD
VWRI525-0636 VWRI525-0637
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Tubes Centrifuge Tubes
Made of high quality, USP class 6 gamma-resistent clear polypropylene with HDPE screw cap.
  • Longer length screw caps with sealing ring prevent leakage
  • Easy-to-read black graduations are accurate to ±2%, with 1 ml increments (15 ml) or 2,5 ml increments (50 ml)
  • Large unerasable frosted white writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Engraved graduation at the conical bottom of each tube
  • Autoclavable at 121 °C and freezable to −80 °C
  • Leakproof
  • During the freezing process, it is better to use plastic rack
  • During high-speed
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M