VWR®, Centrifuge Tubes
Supplier: VWR Collection
VWRI525-0638EA 118 SGD
VWRI525-0638 VWRI525-0639 VWRI525-0640
VWR®, Centrifuge Tubes
Tubes Centrifuge Tubes
PS, clear, conical, with flat cap with integral sealing.
  • Electron beam irradiated
  • Longer length screw caps with sealing ring prevent any leakage
  • Engraved graduations at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M