VWR® Adapt-A Rack®, Test Tube Racks
Supplier: VWR Collection
VWRI525-0702EA 33.6 SGD
VWRI525-0702 VWRI525-0703 VWRI525-0704 VWRI525-0705 VWRI525-0706 VWRI525-0707 VWRI525-0708 VWRI525-0709
VWR® Adapt-A Rack®, Test Tube Racks
Racks Tube Racks
POM.
  • Capable of holding tubes from 12 to 30 mm (approx. 5 to 50 ml) in the same well
  • The slot can hold round, flat, conical and even skirted tubes
  • Link together to create as large or as small a rack as needed
  • A large range of colours available for end user, application or department identification
  • Open sides allow easy label viewing
An innovative flexible multi-tube rack that is capable of holding different sized tubes in each available slot, as well as literally adapting to your tube size to hold firmly in the upright position. Each rackMore Product Information