VWR®, Racks for Centrifuge Tubes
Supplier: VWR Collection
VWRI525-0971EA 67.4 SGD
VWRI525-0971
VWR®, Racks for Centrifuge Tubes
Racks Tube Racks
Rack, PP, white, desgned specifically for 5 ml centrifuge tubes, including Eppendorf brand. It also accommodates other tubes with a diameter of 13 to 16 mm.
  • Durable and corrosion resistant
  • End plates have handles for easy transport and stacking
  • Top shelf height: 34 mm