VWR®, Racks for Centrifuge Tubes, Floating
Supplier: VWR Collection
VWRI525-0972EA 55.2 SGD
VWRI525-0972
VWR®, Racks for Centrifuge Tubes, Floating
Racks Tube Racks
Rack, PP, white. This 'open style' floating rack provides tubes with maximum contact with the water and better circulation of the water around the tubes, allowing superior temperature control of samples.
  • Durable and corrosion resistant
  • Easy transport and stacking
  • Works on a bench top and floats in a water bath
  • Steam autoclavable at 121 °C
Tubes are held tightly in rack to ensure no 'pop-up' tubes or 'floaters', while pressing the legs of the rack on the bench tubes will raise allowing for easy removal.