VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
VWRI525-1066EA 157 SGD
VWRI525-1066 VWRI525-1067 VWRI525-1068 VWRI525-1069 VWRI525-1070 VWRI525-1071 VWRI525-1072 VWRI525-1073 VWRI525-1074 VWRI525-1075 VWRI525-1076 VWRI525-1077 VWRI525-1078 VWRI525-1079 VWRI525-1080
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Tubes Centrifuge Tubes
These leakproof disposable conical bottom and free-standing tubes feature black graduations and smooth inner walls for easy filling and sample preparation.
  • Easy-to-read black graduations in ±2% increments
  • White printed writing area for permanent coding
  • RNase-, DNase-, and endotoxin-free
  • Autoclavable (121 °C) and freezable (–80 °C)
  • Can be centrifuged up to 12500×g
  • Sterile tubes are sterilized by irradiation
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M