VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWRI525-1107EA 474 SGD
VWRI525-1107 VWRI525-1109 VWRI525-1111 VWRI525-1113 VWRI525-1103 VWRI525-1105 VWRI525-1097 VWRI525-1098 VWRI525-1099 VWRI525-1100 VWRI525-1101 VWRI525-1081 VWRI525-1082 VWRI525-1083 VWRI525-1084 VWRI525-1085 VWRI525-1086 VWRI525-1087 VWRI525-1088 VWRI525-1089 VWRI525-1090 VWRI525-1091 VWRI525-1092 VWRI525-1093 VWRI525-1094 VWRI525-1095 VWRI525-1096
VWR® Ultra-High Performance Centrifuge Tubes
Tubes Centrifuge Tubes
These leakproof, conical bottom centrifuge tubes prevent breakage and leakage during high speed centrifugation, and are made of non cytotoxic, medical grade resin that allows users to easily see sample volume and colour.
  • Easy-to-read black graduations in ±2% increments
  • Contains a large, white frosted writing area
  • RNase-, DNase-, and non-pyrogenic
  • Autoclavable at 121 °C and freezable to –80 °C
  • Sterile tubes are sterilized by irradiation
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M