VWR®, Centrifuge Tubes, Borosilicate Glass 3.3
Supplier: VWR Collection
VWRI525-1578EA 13.1 SGD
VWRI525-1578 VWRI525-1579 VWRI525-1580 VWRI525-1581 VWRI525-1582 VWRI525-1583 VWRI525-1584 VWRI525-1585
VWR®, Centrifuge Tubes, Borosilicate Glass 3.3
Tubes Centrifuge Tubes
Reusable centrifuge tubes made from borosilicate 3.3 glass, available with round or conical bottom, graduated or non graduated.
  • Produced from borosilicate 3.3 glass tubing
  • Highly resistant against chemicals and rapid temperature changes
  • Autoclavable
  • Withstand forces up to 3500 ×g
Graduations and inscriptions in high contrasting blue enamel. With straight rim.
Certifications: DIN 58970 (only round bottom tubes).
Order Now
SPECIFICATIONS
Environmentally Preferable