VWR®, Centrifuge Tubes, Heavy Wall, with Screw Cap
Supplier: VWR Collection
VWRI525-1587EA 54.9 SGD
VWRI525-1587 VWRI525-1588
VWR®, Centrifuge Tubes, Heavy Wall, with Screw Cap
Tubes Centrifuge Tubes
Made from borosilicate 3.3 glass, with heavy wall to withstand higher speeds in centrifugation.
  • Produced from borosilicate 3.3 glass tubing
  • Highly resistant against chemicals and rapid temperature changes
  • Autoclavable
  • Withstand forces up to 3500 ×g
  • Thread size 24-410, screw caps included
VWR® heavy wall centrifuge tubes are especially used for separating neutralised sediment for culturing from sputum specimens.