VWR® Mega Racks® Tube Racks for 28 to 32 mm Tubes
Supplier: VWR Collection
VWRI525-1619EA 193 SGD
VWRI525-1619 VWRI525-1620
VWR® Mega Racks® Tube Racks for 28 to 32 mm Tubes
Racks Tube Racks
High-capacity storage for 50 ml tubes.
  • Sturdy and lightweight for easy and secure transport of tubes
  • Place different test groupings into one rack and eliminate need for multiple tube racks
  • Alphanumeric identification for quick location and retrieval of tubes
  • Autoclavable
These sturdy and lightweight mega tube racks are great for high throughput laboratories requiring large sample storage. They allow you to place different test groupings into one rack, eliminating the need for multiple tube racks. These racks also feature alphanumeric identificationMore Product Information