VWR®, Inserts for ND13 Screw Neck Vials
Supplier: VWR Collection
VWRI548-0093AEA 514 SGD
VWRI548-0093A
VWR®, Inserts for ND13 Screw Neck Vials
Vials
ND13 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
ND13 screw neck vials are constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear versions with a flat bottom to ensure compatibility with inserts. Associated screw caps, mostly available with assembled septa, are engineered to prevent sampleMore Product Information