VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWRI548-9006AEA 659 SGD
VWRI548-9006A VWRI548-0423A VWRI548-0172A VWRI548-2500A VWRI548-0905A VWRI548-3311A VWRI548-1861A VWRI548-0467A VWRI548-0173A VWRI548-0174A VWRI548-1858A VWRI548-0003A VWRI548-0447A VWRI548-0004A VWRI548-0005A VWRI548-3337A VWRI548-3310A VWRI548-8008A VWRI548-8009A VWRI548-8010A VWRI548-2380A VWRI548-2148A VWRI548-1361A VWRI548-0171A VWRI548-2159A VWRI548-1362A VWRI548-0856A VWRI548-0474A VWRI548-1465A VWRI548-1364A VWRI548-1400A VWRI548-0906A VWRI548-3254A VWRI548-0092A VWRI548-2870A VWRI548-1516A VWRI548-2126A VWRI548-0418A VWRI548-0421A VWRI548-1840A VWRI548-2146A VWRI548-2362A VWRI548-2512A VWRI548-2603A VWRI548-2604A
VWR®, Crimp Neck Vials ND11
Vials
ND11 crimp neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of aluminium seal styles and septa materials.
  • High performance caps/septa
  • Crimp top vials provide an excellent seal
  • Larger opening improves sample accessibility
ND11 crimp neck vials are available in amber or clear version. For use in a broad variety of instruments as GC and HPLC standard vials with a flat bottom. Multiple crimp cap colours mostly available with assembled stopper or septa are engineered toMore Product Information