VWR®, 96-Position Block Systems
Supplier: VWR Collection
VWRI548-0343EA 874 SGD
VWRI548-0343
VWR®, 96-Position Block Systems
Microplates Microplates with Glass Inserts
96-position systems (Ritter) with glass inserts are used in HTS (high throughput screening) chemistry when the pure PP block is not inert enough. The inserts can be sealed with a PE cap seal, or over the whole block with a Sealmat block cover made of silicone, laminated with a PTFE film.