VWR®, Crimp Caps for Crimp Neck Vials ND13
Supplier: VWR Collection
VWRI548-3313AEA 1060 SGD
VWRI548-3313A VWRI548-1843A VWRI548-0488A VWRI548-0513A VWRI548-0492A VWRI548-1459A VWRI548-0498A VWRI548-2132A VWRI548-0495A VWRI548-0489A VWRI548-2084A VWRI548-1522A VWRI548-0858A VWRI548-0491A VWRI548-0496A
VWR®, Crimp Caps for Crimp Neck Vials ND13
Closures Chromatography Vial Closures
Crimp Neck ND13 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of aluminium seal styles and septa materials.
  • High performance caps/septa
  • Crimp top vials provide an excellent seal
  • Larger opening improves sample accessibility
ND13 vials with a crimp neck are available in clear version. For use in a broad range of instruments as GC and HPLC standard vials with a flat bottom. Broad variety of crimp caps mostly available with assembled stopper or septa are engineered to preventMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable