VWR® ND20, Headspace Crimp Vials
Supplier: VWR Collection
VWRI548-0609AEA 129 SGD
VWRI548-0609A
VWR® ND20, Headspace Crimp Vials
Vials
Clear and amber glass vials with a rounded or flat bottom, and volumes from 5 to 20 ml and closures. A rounded bottom vial is more sturdy and resistant to high pressure during the heating process. Flat bottom vials are more appropriate for certain instruments.
  • Pressure-release safety caps with a broad range of different septa available
  • Magnetic and magnetic bi-metal caps for CTC Combi PAL and Shimadzu AOC5000 autosamplers
  • Caps with Pharma-Fix septa (butyl/PTFE) achieve the best sealing properties
  • Due to the 1,2 mm wall thickness, these vials can withstand
More Product Information