VWR® Screw Neck Vials, ND40
Supplier: VWR Collection
VWRI548-0660AEA 289 SGD
VWRI548-0660A
VWR® Screw Neck Vials, ND40
Vials
Cylindrical vial made of clear glass, 3rd hydrol. class.
Thread DIN40.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable