VWR®, Storage Boxes for Sample Vials/Bottles
Supplier: VWR Collection
VWRI548-1487AEA 291 SGD
VWRI548-1487A
VWR®, Storage Boxes for Sample Vials/Bottles
Boxes Boxes for Vials
Storage boxes for convenient, safe storage and transportation of sample vials and sample bottles. A selection of boxes to securely hold a wide range of vial sizes. Ideal for space-saving storage in fridges, as the transparent lid prevents condensation on the closures so avoiding contamination in the cooling unit.
  • Temperature resistant from −80 to 100 °C
  • Alphanumeric coding (1,5 and 4 ml) for clear sample identification
  • Chemically resistant, fairly robust and stackable
  • PP, coloured
Order Now
SPECIFICATIONS
Environmentally Preferable