VWR® WebSeal™, 96-Well Microplates, PP, Certified
Supplier: VWR Collection
VWRI548-3371EA 255 SGD
VWRI548-3371
VWR® WebSeal™, 96-Well Microplates, PP, Certified
Microplates HTS Microplates
PP, non coated, non sterile, chromatography tested and certified. Manufactured from ultra low bleed high purity basis resin and fully lot tested by GC/MS for organic extractables. Designed for sensitive applications, lower sample concentrations, regulated laboratories, and/or TIC and full chromatograms.
  • Microplates are chemically and thermally resistant and will tolerate temperatures from −80 to +121 °C
  • SBS and ANSI standard footprint design for broad instrument compatibility
  • Ideal for pharmaceutical applications, sample collection and storage,
More Product Information