VWR® PFAS Kit
Supplier: VWR Collection
VWRI548-7001AEA 95.4 SGD
VWRI548-7001A
VWR® PFAS Kit
Vials
Vials, PP, 1,5 ml with 9 mm Short thread cap, Silicone/Polypropylene.
  • For the PFAS sample process
  • Non certified
It is essential during the PFAS sample process to provide vials and caps that are PFAS free. Using the new PFAS vials and caps of VWR® ensure the integrity of the data collected and meets all required collection standards. VWR® has created a series of vials and caps that are compatible with all common autosamplers. These vials and caps are recommended for environmental monitoring, food and water quality testing, product testing, industrial
More Product Information