VWR® ND24, EPA Screw Neck Vials
Supplier: VWR Collection
VWRI548-9025EA 4830 SGD
VWRI548-9025
VWR® ND24, EPA Screw Neck Vials
Vials
These vials are for use on instruments from the following manufacturers: Agilent, Dionex, Shimadzu, Tekmar, Thermo Scientific, Varian.
  • EPA vials can be delivered with a certificate of cleanliness that might be needed for TOC analysis - surcharges apply
  • Can be ordered with any type of ND24 screw seal already screwed on
Order Now
SPECIFICATIONS
Environmentally Preferable