VWR® Standard Micro Syringes
Supplier: VWR Collection
VWRI549-0199EA 74.2 SGD
VWRI549-0199 VWRI549-0522 VWRI549-0523 VWRI549-0524
VWR® Standard Micro Syringes
Syringes Syringes for Chromatography
Standard micro syringes for GC with fixed needle.
  • All parts in contact with liquid are made of stainless steel and neutral borosilicate glass
Length in ordering table refers to needle length.
Order Now
SPECIFICATIONS
Environmentally Preferable