VWR® Gas-Tight Micro Syringes
Supplier: VWR Collection
VWRI549-0539EA 177 SGD
VWRI549-0539 VWRI549-0536 VWRI549-0538 VWRI549-0559 VWRI549-0561 VWRI549-0562 VWRI549-0563 VWRI549-0564 VWRI549-0537
VWR® Gas-Tight Micro Syringes
Syringes Syringes for Chromatography
Micro syringes without needle
General purpose Luer lock connector for use with autosamplers and liquid handling instruments. All parts in contact with sample are made from neutral borosilicate glass and PTFE. This provides excellent performance and an inert sample environment.
Interchangeable needles
549-0560 made from SUS304; 549-0565 made form chrome plated brass. 549-0560 needles for 1; 2,5; 5 and 10 ml gas-tight syringes. 549-0565 needles for 0,1 ml gas-tight syringes.
PTFE tips
Provides a complete leak-tight seal for both liquids and gases.
549-0560 needles for 1, 2,5, 5 and 10 mlMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable