VWR®, TLC Glass Plates, Silica, Unmodified Layers
Supplier: VWR Collection
VWRI551-0006EA 153 SGD
VWRI551-0006 VWRI551-0013 VWRI551-0014 VWRI551-0017 VWRI551-0024 VWRI551-0031
VWR®, TLC Glass Plates, Silica, Unmodified Layers
TLC Plates
These TLC glass plates are suitable for easy and quick sample applications.
  • High sample throughput in short time
  • Pilot product for HPLC or Flash chromatography
  • Suitable for screening tests
Indicator: Manganese activated zinc silica with green fluorescence for short wave UV (254 nm).
Binder: Stable in almost all organic solvents.
Particle size: 5 to 17 µm
Order Now
SPECIFICATIONS
Environmentally Preferable