VWR®, HPTLC Glass Plates, Silica HP, Unmodified Layers

Supplier: VWR Collection
VWRI551-0009EA 454 SGD
VWRI551-0009 VWRI551-0023 VWRI551-0027 VWRI551-0028
VWR®, HPTLC Glass Plates, Silica HP, Unmodified Layers
TLC Plates
These HPTLC glass plates have an excellent separation efficiency due to an optimised particle size distribution.

  • Specific dyeability and abrasion resistance based on an optimised binder system
  • Good wettability for exact colorisation results, even with 100% aqueous detection reagents

Specific surface (BET): ~500 m²/g

Specific pore volume: 0,75 ml/g

Particle size: 2 to 10 µm
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR