VWR® Air Valve
Supplier: VWR Collection
VWRI590-0025EA 29 SGD
VWRI590-0025
VWR® Air Valve
Valves Air Regulation Valves
Valve housing with connection made of black PP (thread UNF 1/4" 28G) suitable for VWR safety caps.
  • 6 month service life
With integrated check valve and upstream membrane made of porous PTFE (1 μm), including labelling field.
Order Now
SPECIFICATIONS
Environmentally Preferable