VWR®, Digital Stopwatch
Supplier: VWR Collection
VWRI609-0054EA 45.4 SGD
VWRI609-0054
VWR®, Digital Stopwatch
Timers Digital Stopwatches
O-ring seal and shockproof plastic case for lab and plant use.
  • Timing capacity: 23 hours, 59 minutes, 59 seconds (count up)
  • LCD shows time of day in hours, minutes and seconds
  • AM/PM, date and day of the week
  • Cumulative split timing
  • Audible alarm sounds for 1 minute
  • Waterproof to a pressure of three atmospheres
Order Now
SPECIFICATIONS
Environmentally Preferable